Product Details

SNP ID
rs13268726
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:125000892 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGCTTTTTAAATTTTTGCGCCACC[A/G]TTTCTGGATGTGAGATCTTTGGCAA
Phenotype
MIM: 602019
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC105375744 PubMed Links

Gene Details

Gene
LOC105375744
Gene Name
uncharacterized LOC105375744
There are no transcripts associated with this gene.

Gene
SQLE
Gene Name
squalene epoxidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003129.3 Intron NP_003120.2
XM_011517246.2 Intron XP_011515548.1

View Full Product Details