Product Details

SNP ID
rs10056639
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:133006490 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTCTTACTAGCTGTATGACAATGA[A/G]TAAGTTATTCAACATCTCCTTGCTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZCCHC10 PubMed Links

Gene Details

Gene
ZCCHC10
Gene Name
zinc finger CCHC-type containing 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300816.2 Intron NP_001287745.1
NM_001300817.2 Intron NP_001287746.1
NM_001300818.2 Intron NP_001287747.1
NM_001300819.2 Intron NP_001287748.1
NM_001300822.2 Intron NP_001287751.1
NM_001308127.1 Intron NP_001295056.1
NM_001308128.1 Intron NP_001295057.1
NM_001308129.1 Intron NP_001295058.1
NM_001308130.1 Intron NP_001295059.1
NM_017665.3 Intron NP_060135.1
XM_011543502.1 Intron XP_011541804.1
XM_011543503.2 Intron XP_011541805.1

View Full Product Details