Product Details

SNP ID
rs2278233
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:17406365 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGGGCCAGCCTCAGACACTGTCCA[C/T]GTGGCCAAGAGAGCCAGGGCTTCCT
Phenotype
MIM: 600534
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
BISPR PubMed Links

Gene Details

Gene
BISPR
Gene Name
BST2 interferon stimulated positive regulator (non-protein coding)
There are no transcripts associated with this gene.

Gene
BST2
Gene Name
bone marrow stromal cell antigen 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004335.3 Intron NP_004326.1
Gene
MVB12A
Gene Name
multivesicular body subunit 12A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304547.1 Intron NP_001291476.1
NM_138401.3 Intron NP_612410.1

View Full Product Details