Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001042544.1 | Intron | NP_001036009.1 | |||
NM_001042545.1 | Intron | NP_001036010.1 | |||
NM_003573.2 | Intron | NP_003564.2 | |||
XM_011527376.2 | Intron | XP_011525678.1 | |||
XM_011527377.2 | Intron | XP_011525679.1 | |||
XM_011527378.2 | Intron | XP_011525680.1 | |||
XM_011527379.1 | Intron | XP_011525681.1 | |||
XM_011527380.2 | Intron | XP_011525682.1 | |||
XM_011527381.2 | Intron | XP_011525683.1 | |||
XM_011527382.2 | Intron | XP_011525684.1 | |||
XM_011527383.2 | Intron | XP_011525685.1 | |||
XM_011527384.2 | Intron | XP_011525686.1 | |||
XM_011527385.2 | Intron | XP_011525687.1 | |||
XM_011527386.2 | Intron | XP_011525688.1 | |||
XM_011527387.1 | Intron | XP_011525689.1 | |||
XM_017027352.1 | Intron | XP_016882841.1 | |||
XM_017027353.1 | Intron | XP_016882842.1 | |||
XM_017027354.1 | Intron | XP_016882843.1 |