Product Details

SNP ID
rs3822426
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:38476426 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATTCTTAACGGAAGTACACTTTA[A/T]ATGTATAAGGAAATTCAACAACTTA
Phenotype
MIM: 151443
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LIFR PubMed Links

Gene Details

Gene
LIFR
Gene Name
leukemia inhibitory factor receptor alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127671.1 8640 UTR 3 NP_001121143.1
NM_002310.5 8640 UTR 3 NP_002301.1
XM_011514040.2 8640 Intron XP_011512342.1
XM_011514042.2 8640 Intron XP_011512344.1
XM_017009462.1 8640 UTR 3 XP_016864951.1
XM_017009463.1 8640 Intron XP_016864952.1

View Full Product Details