Product Details

SNP ID
rs4808621
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:17303458 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCCCGAGTAGGCGGAGCCACGTG[C/T]CGACGGGCGGGCACTTCGGCCTATC
Phenotype
MIM: 611840
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ABHD8 PubMed Links

Gene Details

Gene
ABHD8
Gene Name
abhydrolase domain containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024527.4 16 UTR 5 NP_078803.4
Gene
DDA1
Gene Name
DET1 and DDB1 associated 1
There are no transcripts associated with this gene.

Gene
MRPL34
Gene Name
mitochondrial ribosomal protein L34
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_023937.3 16 Intron NP_076426.1
XM_011528189.2 16 Intron XP_011526491.1

View Full Product Details