Product Details
- SNP ID
-
rs298104
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:52424720 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TTTCCACCTCCAATGCTGCTGCCGC[G/T]TCCATAGCCACTGGAAAGAGTAGAG
- Phenotype
-
MIM: 609025
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
KRT75
PubMed Links
Gene Details
- Gene
- KRT75
- Gene Name
- keratin 75
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004693.2 |
1475 |
Missense Mutation |
|
|
NP_004684.2 |
View Full Product Details