Product Details
- SNP ID
-
rs35258126
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:52459396 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATAGGTTGCCGGTGACAGAATGCGT[C/T]GTCAGACTTTAAGAAGGGCACTTAG
- Phenotype
-
MIM: 615507
MIM: 191040
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
NISCH
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112833562] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NISCH
- Gene Name
- nischarin
- Gene
- TNNC1
- Gene Name
- troponin C1, slow skeletal and cardiac type
There are no transcripts associated with this gene.
View Full Product Details