Product Details

SNP ID
rs34683543
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:58442675 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAAGCCTTGCATATCTGAGACAAA[A/C]CAAGTAGAGTTTGTCTCATAAGTGG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HSF5 PubMed Links

Gene Details

Gene
HSF5
Gene Name
heat shock transcription factor 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080439.2 Intron NP_001073908.2
XM_011524283.1 Intron XP_011522585.1

View Full Product Details