Product Details

SNP ID
rs1005552
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:36593168 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCCAGGGATTAAACAACTGGATTC[C/T]GTAATTCTGAAGCCACTCTTGCCTT
Phenotype
MIM: 616159 MIM: 612275
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DHRS11 PubMed Links

Gene Details

Gene
DHRS11
Gene Name
dehydrogenase/reductase 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024308.3 Intron NP_077284.2
XM_005257658.2 Intron XP_005257715.1
XM_011525233.2 Intron XP_011523535.1
Gene
GGNBP2
Gene Name
gametogenetin binding protein 2
There are no transcripts associated with this gene.

Gene
MRM1
Gene Name
mitochondrial rRNA methyltransferase 1
There are no transcripts associated with this gene.

View Full Product Details