Product Details

SNP ID
rs17114266
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:28774036 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTTAGCCGGGCTGCTCCGGGTCGG[G/T]TGAGGCACTGGGTGCGTGTGGGAGT
Phenotype
MIM: 164874
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FOXG1 PubMed Links

Gene Details

Gene
FOXG1
Gene Name
forkhead box G1
There are no transcripts associated with this gene.

Gene
FOXG1-AS1
Gene Name
FOXG1 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

Gene
LINC01551
Gene Name
long intergenic non-protein coding RNA 1551
There are no transcripts associated with this gene.

Gene
LOC105370424
Gene Name
uncharacterized LOC105370424
There are no transcripts associated with this gene.

View Full Product Details