Product Details

SNP ID
rs2857101
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:32826899 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGTAGTTGGGAGATACAGGAATTA[C/T]TATTCCTGTTTTATGAATAAAGGAC
Phenotype
MIM: 600629 MIM: 170261
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HLA-DOB PubMed Links

Gene Details

Gene
HLA-DOB
Gene Name
major histocompatibility complex, class II, DO beta
There are no transcripts associated with this gene.

Gene
TAP2
Gene Name
transporter 2, ATP binding cassette subfamily B member
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000544.3 4243 UTR 3 NP_000535.3
NM_001290043.1 4243 UTR 3 NP_001276972.1
NM_018833.2 4243 Intron NP_061313.2

View Full Product Details