Product Details

SNP ID
rs2041386
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:6453372 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGACAGCCCAGGTCCCGATTACAGC[C/T]ACACATACTGCCTCCCGTTGGCCCT
Phenotype
MIM: 186711 MIM: 607081 MIM: 185880
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CD27 PubMed Links

Gene Details

Gene
CD27
Gene Name
CD27 molecule
There are no transcripts associated with this gene.

Gene
CD27-AS1
Gene Name
CD27 antisense RNA 1
There are no transcripts associated with this gene.

Gene
TAPBPL
Gene Name
TAP binding protein like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018009.4 Intron NP_060479.3
XM_005253700.3 Intron XP_005253757.1
XM_017019545.1 Intron XP_016875034.1
XM_017019546.1 Intron XP_016875035.1
Gene
VAMP1
Gene Name
vesicle associated membrane protein 1
There are no transcripts associated with this gene.

View Full Product Details