Product Details

SNP ID
rs12856956
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:123988009 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCATACAGGCATATACATGTGGACA[A/G]AGGCTGGGAAGGAAAATGAAAATAG
Phenotype
MIM: 300826
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
STAG2 PubMed Links

Gene Details

Gene
STAG2
Gene Name
stromal antigen 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042749.2 Intron NP_001036214.1
NM_001042750.1 Intron NP_001036215.1
NM_001042751.1 Intron NP_001036216.1
NM_001282418.1 Intron NP_001269347.1
NM_006603.4 Intron NP_006594.3
XM_005262357.2 Intron XP_005262414.1
XM_005262358.2 Intron XP_005262415.1
XM_005262359.3 Intron XP_005262416.1
XM_005262360.2 Intron XP_005262417.1
XM_005262361.2 Intron XP_005262418.1
XM_006724727.1 Intron XP_006724790.1
XM_006724728.3 Intron XP_006724791.1
XM_011531253.2 Intron XP_011529555.1
XM_017029232.1 Intron XP_016884721.1
XM_017029233.1 Intron XP_016884722.1
XM_017029234.1 Intron XP_016884723.1
XM_017029235.1 Intron XP_016884724.1

View Full Product Details