Product Details
- SNP ID
-
rs3130250
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:29657224 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- GAACAGTAGAGATGGCAAGCTTATC[A/G]AGACCCTCTCTGCCCAGCTGCCTCT
- Phenotype
-
MIM: 159465
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MOG
PubMed Links
Gene Details
- Gene
- MOG
- Gene Name
- myelin oligodendrocyte glycoprotein
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001008228.2 |
244 |
Silent Mutation |
TCA,TCG |
S5S |
NP_001008229.1 |
NM_001008229.2 |
244 |
Silent Mutation |
TCA,TCG |
S5S |
NP_001008230.1 |
NM_001170418.1 |
244 |
Silent Mutation |
TCA,TCG |
S5S |
NP_001163889.1 |
NM_002433.4 |
244 |
Silent Mutation |
TCA,TCG |
S5S |
NP_002424.3 |
NM_206809.3 |
244 |
Silent Mutation |
TCA,TCG |
S5S |
NP_996532.2 |
NM_206810.3 |
244 |
Silent Mutation |
TCA,TCG |
S5S |
NP_996533.2 |
NM_206811.3 |
244 |
Silent Mutation |
TCA,TCG |
S5S |
NP_996534.2 |
NM_206812.3 |
244 |
Silent Mutation |
TCA,TCG |
S5S |
NP_996535.2 |
NM_206814.5 |
244 |
Silent Mutation |
TCA,TCG |
S5S |
NP_996537.3 |
XM_005249131.3 |
244 |
Silent Mutation |
TCA,TCG |
S5S |
XP_005249188.1 |
View Full Product Details