Product Details

SNP ID
rs4984892
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.16:572325 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TGACCACTGAGGAAATGGGCATTTG[A/G]GGAGTTCTGCAGCCTGACCCAGAAG
Phenotype
MIM: 605754
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC107987233 PubMed Links

Gene Details

Gene
LOC107987233
Gene Name
uncharacterized LOC107987233
There are no transcripts associated with this gene.

Gene
NHLRC4
Gene Name
NHL repeat containing 4
There are no transcripts associated with this gene.

Gene
PIGQ
Gene Name
phosphatidylinositol glycan anchor biosynthesis class Q
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004204.3 Intron NP_004195.2
NM_148920.2 Intron NP_683721.1
Gene
PRR35
Gene Name
proline rich 35
There are no transcripts associated with this gene.

View Full Product Details