Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_002237.3 | 2309 | UTR 3 | NP_002228.2 | ||
XM_006723785.2 | 2309 | UTR 3 | XP_006723848.1 | ||
XM_011528800.1 | 2309 | UTR 3 | XP_011527102.1 | ||
XM_011528801.1 | 2309 | UTR 3 | XP_011527103.1 | ||
XM_011528802.1 | 2309 | UTR 3 | XP_011527104.1 | ||
XM_011528803.2 | 2309 | UTR 3 | XP_011527105.1 | ||
XM_011528804.1 | 2309 | UTR 3 | XP_011527106.1 | ||
XM_011528805.2 | 2309 | UTR 3 | XP_011527107.1 | ||
XM_011528806.1 | 2309 | UTR 3 | XP_011527108.1 |