Product Details

SNP ID
rs1052163
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:43039806 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAGGTGGTGAGGACCGCTCTCCC[C/T]GAGTCTCTCGATCGGCCTGCTCTGG
Phenotype
MIM: 608955
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100506679 PubMed Links

Gene Details

Gene
LOC100506679
Gene Name
uncharacterized LOC100506679
There are no transcripts associated with this gene.

Gene
TTLL1
Gene Name
tubulin tyrosine ligase like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012263.4 1510 Silent Mutation TCA,TCG S414S NP_036395.1
XM_006724225.3 1510 Silent Mutation TCA,TCG S317S XP_006724288.1
XM_011530108.2 1510 Silent Mutation TCA,TCG S376S XP_011528410.1
XM_011530110.2 1510 Silent Mutation TCA,TCG S300S XP_011528412.1
XM_017028751.1 1510 Silent Mutation TCA,TCG S414S XP_016884240.1
XM_017028752.1 1510 Silent Mutation TCA,TCG S414S XP_016884241.1
XM_017028753.1 1510 Silent Mutation TCA,TCG S376S XP_016884242.1

View Full Product Details