Product Details

SNP ID
rs941410
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:2787331 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GAGGGTGATAATTGTAATAATGATA[A/G]TCAGGGATATTCAGGCCTTGGGAAG
Phenotype
MIM: 603419 MIM: 601117
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SGTA PubMed Links

Gene Details

Gene
SGTA
Gene Name
small glutamine rich tetratricopeptide repeat containing alpha
There are no transcripts associated with this gene.

Gene
THOP1
Gene Name
thimet oligopeptidase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003249.4 Intron NP_003240.1
XM_011528228.2 Intron XP_011526530.1

View Full Product Details