Product Details

SNP ID
rs1190227
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.14:102925361 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TTTCCACCCCAGGGCCGCTTGGTTC[A/C]TTCCTTCAGGTTTTCCAAAGCTGCC
Phenotype
MIM: 605799 MIM: 614062
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
AMN PubMed Links

Gene Details

Gene
AMN
Gene Name
amnion associated transmembrane protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030943.3 Intron NP_112205.2
XM_011537202.2 Intron XP_011535504.1
XM_011537203.2 Intron XP_011535505.1
Gene
CDC42BPB
Gene Name
CDC42 binding protein kinase beta
There are no transcripts associated with this gene.

View Full Product Details