Product Details

SNP ID
rs482182
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:43330021 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCCTCCAGGGTGCAGCTGGAGGTC[C/T]GGGAACCTGCGCGTGGGCCCTGGGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00322 PubMed Links

Gene Details

Gene
LINC00322
Gene Name
long intergenic non-protein coding RNA 322
There are no transcripts associated with this gene.

View Full Product Details