Product Details
- SNP ID
-
rs14335
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
56
- Location
-
Chr.1:157575060 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTTCATTGTTTGCACAGTGCCTGTG[C/T]AGCCACTCACAGTTAAACAAACTGA
- Phenotype
-
MIM: 605876
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FCRL4
PubMed Links
Gene Details
- Gene
- FCRL4
- Gene Name
- Fc receptor like 4
View Full Product Details