Product Details

SNP ID
rs628632
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.11:60925747 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GACGCCATGGGAGTCCTAATAGAGA[A/G]AGGAGGAAAACCTTGTCAAATCCAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM109 PubMed Links

Gene Details

Gene
TMEM109
Gene Name
transmembrane protein 109
There are no transcripts associated with this gene.

Gene
TMEM132A
Gene Name
transmembrane protein 132A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017870.3 434 Intron NP_060340.2
NM_178031.2 434 Intron NP_821174.1
XM_017017951.1 434 UTR 5 XP_016873440.1
XM_017017952.1 434 UTR 5 XP_016873441.1

View Full Product Details