Product Details

SNP ID
rs3753014
Assay Type
Validated
NCBI dbSNP Submissions
37
Location
Chr.1:54548922 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
CGCTTTGGAGCTGAGATTAGTTGCT[C/G]AGGCACACGGCAGACCCAAGTGCCC
Phenotype
MIM: 606803
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ACOT11 PubMed Links

Gene Details

Gene
ACOT11
Gene Name
acyl-CoA thioesterase 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015547.3 Intron NP_056362.1
NM_147161.3 Intron NP_671517.1

View Full Product Details