Product Details

SNP ID
rs11577746
Assay Type
Functionally tested
NCBI dbSNP Submissions
33
Location
Chr.1:15735091 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCTGCAGCCCACCCCGGGGCAGG[A/G]CGCTCCTGCCTCCACAGCCCCGGGG
Phenotype
MIM: 609613 MIM: 610817
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PLEKHM2 PubMed Links

Gene Details

Gene
PLEKHM2
Gene Name
pleckstrin homology and RUN domain containing M2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015164.2 4 Intron NP_055979.2
XM_005245790.3 4 Intron XP_005245847.1
XM_005245791.4 4 Intron XP_005245848.1
XM_017000757.1 4 Intron XP_016856246.1
XM_017000758.1 4 Intron XP_016856247.1
Gene
SLC25A34
Gene Name
solute carrier family 25 member 34
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207348.2 4 Intron NP_997231.1
XM_011541292.2 4 UTR 5 XP_011539594.1
XM_011541293.1 4 UTR 5 XP_011539595.1
XM_017001083.1 4 UTR 5 XP_016856572.1
Gene
TMEM82
Gene Name
transmembrane protein 82
There are no transcripts associated with this gene.

View Full Product Details