Product Details

SNP ID
rs34319667
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:28209585 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGCCTATCCTCTTGCTAGCCTTTG[C/G]CAAGCCTTGTTGCAAACCCTTCACA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DNAJC8 PubMed Links

Gene Details

Gene
DNAJC8
Gene Name
DnaJ heat shock protein family (Hsp40) member C8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014280.2 Intron NP_055095.2

View Full Product Details