Product Details

SNP ID
rs686364
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.21:30215475 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CGTTTTTGGGCAACATTCACTATTG[A/G]GTTATAGAAATCTCTGATGATGGCA
Phenotype
MIM: 611231
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLDN8 PubMed Links

Gene Details

Gene
CLDN8
Gene Name
claudin 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_199328.2 677 Missense Mutation CCA,TCA P151S NP_955360.1
Gene
LINC00307
Gene Name
long intergenic non-protein coding RNA 307
There are no transcripts associated with this gene.

View Full Product Details