Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001201572.1 | 1975 | Intron | NP_001188501.1 | ||
NM_001201584.1 | 1975 | Intron | NP_001188513.1 | ||
NM_004206.3 | 1975 | Intron | NP_004197.1 | ||
NM_032970.3 | 1975 | UTR 3 | NP_116752.1 | ||
XM_005265533.1 | 1975 | UTR 3 | XP_005265590.1 | ||
XM_005265534.1 | 1975 | UTR 3 | XP_005265591.1 | ||
XM_005265535.1 | 1975 | UTR 3 | XP_005265592.1 | ||
XM_011534212.1 | 1975 | UTR 3 | XP_011532514.1 | ||
XM_017007438.1 | 1975 | Intron | XP_016862927.1 | ||
XM_017007439.1 | 1975 | Intron | XP_016862928.1 |