Product Details
- SNP ID
-
rs4897157
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:125781835 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGTATCTCCCAGAAATAATCTATGC[A/T]CATACAACACATATCTGGTTCATTT
- Phenotype
-
MIM: 609752
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
NCOA7
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78930336] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NCOA7
- Gene Name
- nuclear receptor coactivator 7
View Full Product Details