Product Details
- SNP ID
-
rs11076120
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:55846913 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAGCCCCGGGTGCCTTGTATTTGAT[G/T]AATCCTTTTCACACCCCTCCTCCCA
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CES5A
PubMed Links
Gene Details
- Gene
- CES5A
- Gene Name
- carboxylesterase 5A
View Full Product Details