Product Details

SNP ID
rs8102140
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:17512530 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACGAGGAATCAGGTGTAGAGCCTGA[A/G]TCTCAGGGTCCTGATCTCTAACCCC
Phenotype
MIM: 604951 MIM: 600691
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PGLS PubMed Links

Gene Details

Gene
PGLS
Gene Name
6-phosphogluconolactonase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012088.2 500 Intron NP_036220.1
XM_011527897.2 500 UTR 5 XP_011526199.1
Gene
SLC27A1
Gene Name
solute carrier family 27 member 1
There are no transcripts associated with this gene.

View Full Product Details