Product Details

SNP ID
rs35735569
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:16143787 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
AAGGGTCTCCCAGGTATGTGACCCA[A/G]GAGCCTCAGCCCTCGAGGAGACAGA
Phenotype
MIM: 608349
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HSH2D PubMed Links

Gene Details

Gene
HSH2D
Gene Name
hematopoietic SH2 domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291274.1 Intron NP_001278203.1
NM_032855.3 Intron NP_116244.1
XM_011528386.2 Intron XP_011526688.1
XM_011528387.2 Intron XP_011526689.1
XM_011528388.2 Intron XP_011526690.1
XM_011528389.2 Intron XP_011526691.1
XM_011528390.1 Intron XP_011526692.1
XM_017027402.1 Intron XP_016882891.1
XM_017027403.1 Intron XP_016882892.1
XM_017027404.1 Intron XP_016882893.1

View Full Product Details