Product Details

SNP ID
rs11076716
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:88814557 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATGAAAATTGAGAAAAAGAACATG[C/G]AGTTATTCAAGCTCTTCTGGACCCA
Phenotype
MIM: 102600 MIM: 605525 MIM: 612222
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
APRT PubMed Links

Gene Details

Gene
APRT
Gene Name
adenine phosphoribosyltransferase
There are no transcripts associated with this gene.

Gene
CDT1
Gene Name
chromatin licensing and DNA replication factor 1
There are no transcripts associated with this gene.

Gene
GALNS
Gene Name
galactosamine (N-acetyl)-6-sulfatase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000512.4 4478 Intron NP_000503.1
NM_001323543.1 4478 Intron NP_001310472.1
NM_001323544.1 4478 Intron NP_001310473.1
XM_005256301.3 4478 UTR 3 XP_005256358.1
XM_011522982.2 4478 UTR 3 XP_011521284.1
XM_017023111.1 4478 Intron XP_016878600.1
XM_017023112.1 4478 UTR 3 XP_016878601.1
XM_017023113.1 4478 UTR 3 XP_016878602.1
Gene
LOC107987238
Gene Name
proline-rich proteoglycan 2-like
There are no transcripts associated with this gene.

View Full Product Details