Product Details

SNP ID
rs182149
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:64697362 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAAATAAGTAGAAGATGTATTGAC[C/T]AGATTGCAAACTAGTTAAAATATTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM159B PubMed Links

Gene Details

Gene
FAM159B
Gene Name
family with sequence similarity 159 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164442.1 Intron NP_001157914.1

View Full Product Details