Product Details
- SNP ID
-
rs1259842
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:54151997 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACGTGCCAGGCACTGGGCTAGGCTC[G/T]GGGAACATGAGGATGAAAAAGCACA
- Phenotype
-
MIM: 603547
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MBD2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74508605] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MBD2
- Gene Name
- methyl-CpG binding domain protein 2
View Full Product Details