Product Details

SNP ID
rs1842479
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:70240962 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATGTTTTTATCTTGGTGGGAGAAA[A/T]GATTACTAGCACTCTCAGATGTTCT
Phenotype
MIM: 601695 MIM: 607241
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CSN3 PubMed Links

Gene Details

Gene
CSN3
Gene Name
casein kappa
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005212.2 Intron NP_005203.2
XM_011531620.2 Intron XP_011529922.1
XM_017007761.1 Intron XP_016863250.1
Gene
FDCSP
Gene Name
follicular dendritic cell secreted protein
There are no transcripts associated with this gene.

View Full Product Details