Product Details

SNP ID
rs11926927
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:38268005 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAATTAGATGGATGTTTTCAAGG[A/G]TTTGGGACCTCTGTTCTTTGGTGTT
Phenotype
MIM: 604047
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC22A13 PubMed Links

Gene Details

Gene
SLC22A13
Gene Name
solute carrier family 22 member 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004256.3 Intron NP_004247.2
XM_017007519.1 Intron XP_016863008.1

View Full Product Details