Product Details

SNP ID
rs433551
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:32031950 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGACAGAAAATCTCAACTGGACACA[C/T]ATGTTTGTGAGCCGCGGAAGTTTTT
Phenotype
MIM: 614818
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FRY PubMed Links

Gene Details

Gene
FRY
Gene Name
FRY microtubule binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_023037.2 Intron NP_075463.2
XM_006719749.3 Intron XP_006719812.1
XM_017020303.1 Intron XP_016875792.1
XM_017020304.1 Intron XP_016875793.1
XM_017020305.1 Intron XP_016875794.1
XM_017020306.1 Intron XP_016875795.1
XM_017020307.1 Intron XP_016875796.1
Gene
FRY-AS1
Gene Name
FRY antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details