Product Details
- SNP ID
-
rs163443
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:69168407 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTCTTGTCCCTTATTTCACTGATAC[A/G]TGCAAGAGCATTCAAATGAATAGTA
- Phenotype
-
MIM: 123836
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CCNB1
PubMed Links
Gene Details
- Gene
- CCNB1
- Gene Name
- cyclin B1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_031966.3 |
|
Intron |
|
|
NP_114172.1 |
View Full Product Details