Product Details

SNP ID
rs4681189
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:149764187 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAAGAGCTGCACTGGTCATTTAGA[A/G]GGGTGCTCAGACCACCTGACTGCCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKUB1 PubMed Links

Gene Details

Gene
ANKUB1
Gene Name
ankyrin repeat and ubiquitin domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144960.2 Intron NP_001138432.1
NM_001315505.1 Intron NP_001302434.1
NM_001315506.1 Intron NP_001302435.1
XM_011512798.1 Intron XP_011511100.1
XM_011512799.1 Intron XP_011511101.1
XM_011512801.1 Intron XP_011511103.1
XM_011512802.1 Intron XP_011511104.1

View Full Product Details