Product Details

SNP ID
rs26180
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.5:119452501 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GACCCTCGTCCCGCCCCCGCCATTC[C/G]CCGCCTCCTCCTGTCCCGCAGTCGG
Phenotype
MIM: 601860
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HSD17B4 PubMed Links

Gene Details

Gene
HSD17B4
Gene Name
hydroxysteroid 17-beta dehydrogenase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000414.3 59 UTR 5 NP_000405.1
NM_001199291.2 59 UTR 5 NP_001186220.1
NM_001199292.1 59 UTR 5 NP_001186221.1
NM_001292027.1 59 UTR 5 NP_001278956.1
NM_001292028.1 59 UTR 5 NP_001278957.1

View Full Product Details