Product Details
- SNP ID
-
rs1325080
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
35
- Location
-
Chr.6:96020632 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- ATAAGCTCTAAACTGTATTCTTGGT[G/T]ATGGTGGCTCATCACCATCATTTCA
- Phenotype
-
MIM: 606865
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
FUT9
PubMed Links
- Additional Information
-
For this assay, SNP rs9322606 is located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the NCBI SNP database (dbSNP). A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FUT9
- Gene Name
- fucosyltransferase 9
View Full Product Details