Product Details

SNP ID
rs685377
Assay Type
Functionally tested
NCBI dbSNP Submissions
31
Location
Chr.1:111144991 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCTCAAAATGTAAGAAATTAGCTT[G/T]GATTTCTCTCCTTGATTTGAAATAT
Phenotype
MIM: 616751 MIM: 613360
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CEPT1 PubMed Links

Gene Details

Gene
CEPT1
Gene Name
choline/ethanolamine phosphotransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_006710285.3 Intron XP_006710348.1
XM_011540496.1 Intron XP_011538798.1
Gene
DRAM2
Gene Name
DNA damage regulated autophagy modulator 2
There are no transcripts associated with this gene.

View Full Product Details