Product Details
- SNP ID
-
rs9496646
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:143499910 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGGAAAGGAGAGAAATGGGAAGATC[A/G]CTAGAAATGCAAGTGAAATTGAGTT
- Phenotype
-
MIM: 136820
MIM: 603164
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FUCA2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs139851521] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FUCA2
- Gene Name
- fucosidase, alpha-L- 2, plasma
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_032020.4 |
|
Intron |
|
|
NP_114409.2 |
- Gene
- PEX3
- Gene Name
- peroxisomal biogenesis factor 3
There are no transcripts associated with this gene.
View Full Product Details