Product Details

SNP ID
rs1825368
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.6:151454124 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
TTTATGTTGTTCAAGCACATTAGAC[A/C]CTTTAGGTGGTTTTAGATGCATTGC
Phenotype
MIM: 616332 MIM: 614917
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ARMT1 PubMed Links

Gene Details

Gene
ARMT1
Gene Name
acidic residue methyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286562.1 Intron NP_001273491.1
NM_024573.2 Intron NP_078849.1
Gene
RMND1
Gene Name
required for meiotic nuclear division 1 homolog
There are no transcripts associated with this gene.

View Full Product Details