Product Details

SNP ID
rs12873395
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:72759614 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCATAAATAATTCTGTTCAACCCA[C/G]AAGTATAGTAGTATGATGGGTCAGA
Phenotype
MIM: 610510 MIM: 607533
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
BORA PubMed Links

Gene Details

Gene
BORA
Gene Name
bora, aurora kinase A activator
There are no transcripts associated with this gene.

Gene
DIS3
Gene Name
DIS3 homolog, exosome endoribonuclease and 3'-5' exoribonuclease
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128226.2 3354 UTR 3 NP_001121698.1
NM_001322348.1 3354 UTR 3 NP_001309277.1
NM_001322349.1 3354 UTR 3 NP_001309278.1
NM_014953.4 3354 UTR 3 NP_055768.3

View Full Product Details