Product Details

SNP ID
rs2074639
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:840984 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GCATTGGGCTATAAGAGGAGCTTGA[C/T]CGTGGGTGCACCCTGGACCCCACCA
Phenotype
MIM: 162815 MIM: 177020
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AZU1 PubMed Links

Gene Details

Gene
AZU1
Gene Name
azurocidin 1
There are no transcripts associated with this gene.

Gene
PRTN3
Gene Name
proteinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002777.3 20 Intron NP_002768.3
XM_011528136.1 20 UTR 5 XP_011526438.1

View Full Product Details