Product Details

SNP ID
rs9866791
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:112990186 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCTGTTCATTAATTTTTGTATCA[C/T]ATTTATTCAAGAATTGTACTGTTTA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GTPBP8 PubMed Links

Gene Details

Gene
GTPBP8
Gene Name
GTP binding protein 8 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014170.2 Intron NP_054889.2
NM_138485.1 Intron NP_612494.1

View Full Product Details