Product Details

SNP ID
rs3796343
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:52290153 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGACGAGTAAGATCCTGCAGCTGC[A/G]GGCTTCAGGAAGTCTCCTGGGGCTA
Phenotype
MIM: 610516
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GLYCTK PubMed Links

Gene Details

Gene
GLYCTK
Gene Name
glycerate kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144951.1 Intron NP_001138423.1
NM_145262.3 Intron NP_660305.2
XM_017005730.1 Intron XP_016861219.1
Gene
GLYCTK-AS1
Gene Name
GLYCTK antisense RNA 1
There are no transcripts associated with this gene.

Gene
MIR135A1
Gene Name
microRNA 135a-1
There are no transcripts associated with this gene.

View Full Product Details