Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_033449.2 | 1265 | UTR 3 | NP_258260.1 | ||
XM_005268524.4 | 1265 | Intron | XP_005268581.1 | ||
XM_006714803.3 | 1265 | Intron | XP_006714866.1 | ||
XM_011537698.2 | 1265 | Intron | XP_011536000.1 | ||
XM_011537700.2 | 1265 | Intron | XP_011536002.1 | ||
XM_011537701.2 | 1265 | Intron | XP_011536003.1 | ||
XM_017010013.1 | 1265 | Intron | XP_016865502.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001130029.1 | 1265 | Missense Mutation | ATC,CTC | I133L | NP_001123501.1 |
NM_173828.4 | 1265 | Missense Mutation | ATC,CTC | I133L | NP_776189.3 |
XM_005268414.3 | 1265 | Missense Mutation | ATC,CTC | I133L | XP_005268471.1 |
XM_011537624.1 | 1265 | Missense Mutation | ATC,CTC | I133L | XP_011535926.1 |
XM_011537625.2 | 1265 | Missense Mutation | ATC,CTC | I133L | XP_011535927.1 |